Tisk…

Anti-ORC2 Mouse Monoclonal Antibody

Dodavatel: Abgent
ABGEAT3152AEA 9870 CZK
ABGEAT3152A
Anti-ORC2 Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: ORC2 (origin recognition complex, subunit 2)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR