Tisk…
Anti-OXSR1 Mouse Monoclonal Antibody
Dodavatel: Abgent
ABGEAT3162AEA 9870 CZK
ABGEAT3162A
Anti-OXSR1 Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: OXSR1 (oxidative stress responsive 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human