Tisk…

Anti-RPRD1A Mouse Monoclonal Antibody

Dodavatel: Abgent
ABGEAT3166AEA 9870 CZK
ABGEAT3166A
Anti-RPRD1A Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: RPRD1A (regulation of nuclear pre-mRNA domain containing 1A)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR