Tisk…

Anti-PDLIM7 Mouse Monoclonal Antibody

Dodavatel: Abgent
ABGEAT3264AEA 9870 CZK
ABGEAT3264A
Anti-PDLIM7 Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: PDLIM7 (PDZ and LIM domain 7 (enigma))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR