Tisk…

Anti-ACCS Mouse Monoclonal Antibody

Dodavatel: Abgent
ABGEAT3291AEA 9870 CZK
ABGEAT3291A
Anti-ACCS Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: ACCS (1-aminocyclopropane-1-carboxylate synthase homolog (Arabidopsis)(non-functional))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR