Tisk…

Anti-PI3 Mouse Monoclonal Antibody

Dodavatel: Abgent
ABGEAT3305AEA 9870 CZK
ABGEAT3305A
Anti-PI3 Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: PI3 (peptidase inhibitor 3, skin-derived)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR