Tisk…

Anti-PLA2G1B Mouse Monoclonal Antibody

Dodavatel: Abgent
ABGEAT3329AEA 9870 CZK
ABGEAT3329A
Anti-PLA2G1B Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: PLA2G1B (phospholipase A2, group IB (pancreas))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR