Tisk…

Anti-PLEK Mouse Monoclonal Antibody

Dodavatel: Abgent
ABGEAT3339AEA 9870 CZK
ABGEAT3339A
Anti-PLEK Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: PLEK (pleckstrin)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR