Tisk…
Anti-PLTP Mouse Monoclonal Antibody
Dodavatel: Abgent
ABGEAT3351AEA 9870 CZK
ABGEAT3351A
Anti-PLTP Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: PLTP (phospholipid transfer protein)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human