Tisk…
Anti-PLXNC1 Mouse Monoclonal Antibody
Dodavatel: Abgent
ABGEAT3353AEA 9870 CZK
ABGEAT3353A
Anti-PLXNC1 Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: PLXNC1 (plexin C1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human