Tisk…
Anti-POLA1 Mouse Monoclonal Antibody
Dodavatel: Abgent
ABGEAT3365AEA 9870 CZK
ABGEAT3365A
Anti-POLA1 Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: POLA1 (polymerase (DNA directed), alpha 1, catalytic subunit)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human