Tisk…

Anti-POLE2 Mouse Monoclonal Antibody

Dodavatel: Abgent
ABGEAT3369AEA 9870 CZK
ABGEAT3369A
Anti-POLE2 Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: POLE2 (polymerase (DNA directed), epsilon 2, accessory subunit)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR