Tisk…

Anti-POLK Mouse Monoclonal Antibody

Dodavatel: Abgent
ABGEAT3371AEA 9870 CZK
ABGEAT3371A
Anti-POLK Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: POLK (polymerase (DNA directed) kappa)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR