Tisk…

Anti-POLR1C Mouse Monoclonal Antibody

Dodavatel: Abgent
ABGEAT3372AEA 9870 CZK
ABGEAT3372A
Anti-POLR1C Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: POLR1C (polymerase (RNA) I polypeptide C, 30kDa)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR