Tisk…

Anti-PPARGC1A Mouse Monoclonal Antibody

Dodavatel: Abgent
ABGEAT3397AEA 9870 CZK
ABGEAT3397A
Anti-PPARGC1A Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: PPARGC1A (peroxisome proliferator-activated receptor gamma, coactivator 1 alpha)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR