Tisk…
Anti-PRDX5 Mouse Monoclonal Antibody
Dodavatel: Abgent
ABGEAT3424AEA 9870 CZK
ABGEAT3424A
Anti-PRDX5 Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: PRDX5 (peroxiredoxin 5)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human