Tisk…

Anti-PSMA4 Mouse Monoclonal Antibody

Dodavatel: Abgent
ABGEAT3454AEA 9870 CZK
ABGEAT3454A
Anti-PSMA4 Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: PSMA4 (proteasome (prosome, macropain) subunit, alpha type, 4)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR