Tisk…

Anti-PSMB8 Mouse Monoclonal Antibody

Dodavatel: Abgent
ABGEAT3463AEA 9870 CZK
ABGEAT3463A
Anti-PSMB8 Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: PSMB8 (proteasome (prosome, macropain) subunit, beta type, 8)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR