Tisk…

Anti-PSMC3 Mouse Monoclonal Antibody

Dodavatel: Abgent
ABGEAT3465AEA 9870 CZK
ABGEAT3465A
Anti-PSMC3 Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: PSMC3 (proteasome (prosome, macropain) 26S subunit, ATPase, 3)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR