Tisk…

Anti-PSMD14 Mouse Monoclonal Antibody

Dodavatel: Abgent
ABGEAT3469AEA 9870 CZK
ABGEAT3469A
Anti-PSMD14 Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: PSMD14 (proteasome (prosome, macropain) 26S subunit, non-ATPase, 14)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR