Tisk…
Anti-RAP2B Mouse Monoclonal Antibody
Dodavatel: Abgent
ABGEAT3564AEA 9870 CZK
ABGEAT3564A
Anti-RAP2B Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: RAP2B (RAP2B, member of RAS oncogene family)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human