Tisk…

Anti-RCHY1 Mouse Monoclonal Antibody

Dodavatel: Abgent
ABGEAT3605AEA 9870 CZK
ABGEAT3605A
Anti-RCHY1 Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: RCHY1 (ring finger and CHY zinc finger domain containing 1, E3 ubiquitin protein ligase)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR