Tisk…
Anti-RIOK3 Mouse Monoclonal Antibody
Dodavatel: Abgent
ABGEAT3644AEA 9870 CZK
ABGEAT3644A
Anti-RIOK3 Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: RIOK3 (RIO kinase 3)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human