Tisk…
Anti-CENPP Mouse Monoclonal Antibody
Dodavatel: Abgent
ABGEAT3693AEA 9870 CZK
ABGEAT3693A
Anti-CENPP Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: CENPP (centromere protein P)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human