Tisk…

Anti-RUNX1T1 Mouse Monoclonal Antibody

Dodavatel: Abgent
ABGEAT3740AEA 9870 CZK
ABGEAT3740A
Anti-RUNX1T1 Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: RUNX1T1 (runt-related transcription factor 1; translocated to, 1 (cyclin D-related))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR