Tisk…

Anti-SCARA3 Mouse Monoclonal Antibody

Dodavatel: Abgent
ABGEAT3783AEA 9870 CZK
ABGEAT3783A
Anti-SCARA3 Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: SCARA3 (scavenger receptor class A, member 3)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR