Tisk…

Anti-SCARB2 Mouse Monoclonal Antibody

Dodavatel: Abgent
ABGEAT3784AEA 9870 CZK
ABGEAT3784A
Anti-SCARB2 Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: SCARB2 (scavenger receptor class B, member 2)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR