Tisk…

Anti-SEC22B Mouse Monoclonal Antibody

Dodavatel: Abgent
ABGEAT3805AEA 9870 CZK
ABGEAT3805A
Anti-SEC22B Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: SEC22B (SEC22 vesicle trafficking protein homolog B (S. cerevisiae) (gene/pseudogene))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR