Tisk…
Anti-SIX3 Mouse Monoclonal Antibody
Dodavatel: Abgent
ABGEAT3893AEA 9870 CZK
ABGEAT3893A
Anti-SIX3 Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: SIX3 (SIX homeobox 3)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human