Tisk…

Anti-SLC36A2 Mouse Monoclonal Antibody

Dodavatel: Abgent
ABGEAT3923AEA 9870 CZK
ABGEAT3923A
Anti-SLC36A2 Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: SLC36A2 (solute carrier family 36 (proton/amino acid symporter), member 2)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR