Tisk…
Anti-SNAI2 Mouse Monoclonal Antibody
Dodavatel: Abgent
ABGEAT3968AEA 9870 CZK
ABGEAT3968A
Anti-SNAI2 Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: SNAI2 (snail family zinc finger 2)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human