Tisk…

Anti-SNX25 Mouse Monoclonal Antibody

Dodavatel: Abgent
ABGEAT3986AEA 9870 CZK
ABGEAT3986A
Anti-SNX25 Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: SNX25 (sorting nexin 25)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR