Tisk…

Anti-SPARC Mouse Monoclonal Antibody

Dodavatel: Abgent
ABGEAT4014AEA 9870 CZK
ABGEAT4014A
Anti-SPARC Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: SPARC (secreted protein, acidic, cysteine-rich (osteonectin))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR