Tisk…

Anti-ST8SIA2 Mouse Monoclonal Antibody

Dodavatel: Abgent
ABGEAT4053AEA 9870 CZK
ABGEAT4053A
Anti-ST8SIA2 Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: ST8SIA2 (ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 2)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR