Tisk…

Anti-STK38 Mouse Monoclonal Antibody

Dodavatel: Abgent
ABGEAT4086AEA 9870 CZK
ABGEAT4086A
Anti-STK38 Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: STK38 (serine/threonine kinase 38)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR