Tisk…

Anti-TAF1 Mouse Monoclonal Antibody

Dodavatel: Abgent
ABGEAT4137AEA 9870 CZK
ABGEAT4137A
Anti-TAF1 Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: TAF1 (TAF1 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 250kDa)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR