Tisk…

Anti-TAF7 Mouse Monoclonal Antibody

Dodavatel: Abgent
ABGEAT4147AEA 9870 CZK
ABGEAT4147A
Anti-TAF7 Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: TAF7 (TAF7 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 55kDa)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR