Tisk…

Anti-TES Mouse Monoclonal Antibody

Dodavatel: Abgent
ABGEAT4207AEA 9870 CZK
ABGEAT4207A
Anti-TES Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: TES (testis derived transcript (3 LIM domains))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR