Tisk…

Anti-TP53RK Mouse Monoclonal Antibody

Dodavatel: Abgent
ABGEAT4313AEA 9870 CZK
ABGEAT4313A
Anti-TP53RK Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: TP53RK (TP53 regulating kinase)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR