Tisk…
Anti-UNG Mouse Monoclonal Antibody
Dodavatel: Abgent
ABGEAT4473AEA 9870 CZK
ABGEAT4473A
Anti-UNG Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: UNG (uracil-DNA glycosylase)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human