Tisk…
Anti-WDR20 Mouse Monoclonal Antibody
Dodavatel: Abgent
ABGEAT4529AEA 9870 CZK
ABGEAT4529A
Anti-WDR20 Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: WDR20 (WD repeat domain 20)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human