Tisk…
Anti-XBP1 Mouse Monoclonal Antibody
Dodavatel: Abgent
ABGEAT4549AEA 9870 CZK
ABGEAT4549A
Anti-XBP1 Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: XBP1 (X-box binding protein 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human