Tisk…

Anti-CDCA8 Mouse Monoclonal Antibody

Dodavatel: Abgent
ABGEAM1868AEA 0 CZK
ABGEAM1868A
Anti-CDCA8 Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: CDCA8 (cell division cycle associated 8)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR