Tisk…

Anti-Myc Tag Polyclonal Antibody

Dodavatel: OriGene
ORIGTA100023EA 15250 CZK
ORIGTA100023
Anti-Myc Tag Polyclonal Antibody
Protilátky
Type: Tag
Antigen: Myc Tag (Myc tag)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR