Tisk…

Anti-DDK Tag Monoclonal Antibody (DyLight® 488)

Dodavatel: OriGene
ORIGTA150021EA 15000 CZK
ORIGTA150021
Anti-DDK Tag Monoclonal Antibody (DyLight® 488)
Protilátky
Type: Tag
Antigen: DDK Tag
Clonality: monoclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: None
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR