Tisk…

Anti-mCherry Mouse Monoclonal Antibody

Dodavatel: OriGene
ORIGTA180028EA 14010 CZK
ORIGTA180028
Anti-mCherry Mouse Monoclonal Antibody
Protilátky
Type: Tag
Antigen: mCherry
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR