Tisk…

Anti-NAE1 Rabbit Polyclonal Antibody

Dodavatel: OriGene
ORIGTA300061EA 19680 CZK
ORIGTA300061
Anti-NAE1 Rabbit Polyclonal Antibody
Protilátky
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50-100

Type: Primary
Antigen: NAE1 (NEDD8 activating enzyme E1 subunit 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR