Tisk…

Anti-S100B Rabbit Monoclonal Antibody [clone: EP1576Y]

Dodavatel: OriGene
ORIGTA300915EA 20270 CZK
ORIGTA300915
Anti-S100B Rabbit Monoclonal Antibody [clone: EP1576Y]
Recommended Dilutions: Western Blot: 1:1000; Immunoprecipitation: 1:40

Type: Primary
Antigen: S100B (S100 calcium binding protein B)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR