Tisk…

Anti-HMGB1 Rabbit Polyclonal Antibody

Dodavatel: OriGene
ORIGTA301448EA 21030 CZK
ORIGTA301448
Anti-HMGB1 Rabbit Polyclonal Antibody
Protilátky
Recommended Dilutions: Western Blot: 0.5-1.0 ?g/ml

Type: Primary
Antigen: HMGB1 (high mobility group box 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR