Tisk…

Anti-YAP1 Rabbit Polyclonal Antibody

Dodavatel: OriGene
ORIGTA301498EA 21060 CZK
ORIGTA301498
Anti-YAP1 Rabbit Polyclonal Antibody
Protilátky
Recommended Dilutions: Western Blot: 1: 1000

Type: Primary
Antigen: YAP1 (Yes-associated protein 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR